accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NH3COHO

CAS RN 99-64-9

IUPAC Name: 3-(dimethylamino)benzoic acid
Synonyms: 3-(dimethylamino)benzoic acid
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: NEGFNJRAUMCZMY-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=CC(=C1)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results