accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 98534-80-6

NNOOHFFFCl

CAS RN 98534-80-6

IUPAC Name: 1-(4-chlorophenyl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid
Synonyms: 1-(4-chlorophenyl)-5-(trifluoromethyl)pyrazole-4-carboxylic acid
Molecular Weight (g/mol): 290.63
Molecular Formula: C11H6ClF3N2O2
InChi Key: HSZJMDBOSZVDOK-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(N(N=C1)C1=CC=C(Cl)C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results