accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 983-81-3

PP(E)

CAS RN 983-81-3

IUPAC Name: [(1E)-2-(diphenylphosphanyl)ethenyl]diphenylphosphane
Synonyms: [(1E)-2-(diphenylphosphanyl)ethenyl]diphenylphosphane
Molecular Weight (g/mol): 396.41
Molecular Formula: C26H22P2
InChi Key: NCKJIJSEWKIXAT-QURGRASLSA-N
SMILES: C(=C/P(C1=CC=CC=C1)C1=CC=CC=C1)\P(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results