accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 98064-96-1

H3COOOHOOOHOHOHOHOHHO

CAS RN 98064-96-1

IUPAC Name: 2-{[6-(hexadecyloxy)-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms: 2-{[6-(hexadecyloxy)-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol): 566.73
Molecular Formula: C28H54O11
InChi Key: GTQCHJYVKDXMRU-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCCCCCOC1OC(CO)C(OC2OC(CO)C(O)C(O)C2O)C(O)C1O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results