CAS RN 98026-16-5
CAS RN 98026-16-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N,N-Dimethylamidino Urea, TRC
CAS: 98026-16-5 Molecular Formula: C4H10N4O Molecular Weight (g/mol): 130.15 Synonym: N-[(Dimethylamino)iminomethyl]Urea,N3-Desamidino,N3-carbamoyl Metformin,N3-Desamidino,N3-hydroxyimidoyl Metformin,Metformin Hydroxy Analog IUPAC Name: (N,N-dimethylcarbamimidoyl)urea SMILES: NC(NC(N(C)C)=N)=O≫N=C(O)NC(N(C)C)=N