accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3H3CCH3OHO

CAS RN 98-73-7

IUPAC Name: 4-tert-butylbenzoic acid
Synonyms: 4-tert-butylbenzoic acid kyselina p-terc.butylbenzoova 4-T-butylbenzoic acid TBBA
Molecular Weight (g/mol): 178.23
Molecular Formula: C11H14O2
InChi Key: KDVYCTOWXSLNNI-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(C=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results