accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 98-50-0

NH2AsOHHOO

CAS RN 98-50-0

IUPAC Name: (4-aminophenyl)arsonic acid
Synonyms: 4-aminophenylarsonic acid arsanilic acid p-aminophenylarsine acid pro-gen 227 premix acide p-arsanilique atoxylic acid C6H8AsNO3 kyselina arsanilova arsanilic acid-100 antoxylic acid
Molecular Weight (g/mol): 217.06
Molecular Formula: C6H8AsNO3
InChi Key: XKNKHVGWJDPIRJ-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)[As](O)(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)