accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 97888-80-7

CH3H3COOO

CAS RN 97888-80-7

IUPAC Name: 4-(benzyloxy)-3,5-dimethylbenzoate
Synonyms: 4-(benzyloxy)-3,5-dimethylbenzoate
Molecular Weight (g/mol): 255.29
Molecular Formula: C16H15O3
InChi Key: JABUPJCJZZNUFK-UHFFFAOYSA-M
SMILES: CC1=CC(=CC(C)=C1OCC1=CC=CC=C1)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results