accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 97839-99-1

NHOCl

CAS RN 97839-99-1

IUPAC Name: 4-(4-chlorophenoxy)piperidine
Synonyms: 4-(4-chlorophenoxy)piperidine
Molecular Weight (g/mol): 211.69
Molecular Formula: C11H14ClNO
InChi Key: VOQMPZXAFLPTMM-UHFFFAOYSA-N
SMILES: ClC1=CC=C(OC2CCNCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results