accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 97603-94-6

IUPAC Name: {amino[3,5-bis(trifluoromethyl)phenyl]methylidene}azanium
Synonyms: {amino[3,5-bis(trifluoromethyl)phenyl]methylidene}azanium
Molecular Weight (g/mol): 257.16
Molecular Formula: C9H7F6N2
InChi Key: NKSWBGJHZIMJCJ-UHFFFAOYSA-O
SMILES: NC(=[NH2+])C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results