accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3SH3CSOOCl

CAS RN 97272-04-3

IUPAC Name: 2,5-dimethylthiophene-3-sulfonyl chloride
Synonyms: 2,5-dimethylthiophene-3-sulfonyl chloride
Molecular Weight (g/mol): 210.69
Molecular Formula: C6H7ClO2S2
InChi Key: CMTPCYKEUFDVAU-UHFFFAOYSA-N
SMILES: CC1=CC(=C(C)S1)S(Cl)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results