accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 97075-46-2

HNNHHClHCl

CAS RN 97075-46-2

IUPAC Name: (diphenylmethyl)({2-[(diphenylmethyl)amino]ethyl})amine dihydrochloride
Synonyms: (diphenylmethyl)({2-[(diphenylmethyl)amino]ethyl})amine dihydrochloride
Molecular Weight (g/mol): 465.46
Molecular Formula: C28H30Cl2N2
InChi Key: YRQCDCNQANSUPB-UHFFFAOYSA-N
SMILES: Cl.Cl.C(CNC(C1=CC=CC=C1)C1=CC=CC=C1)NC(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results