accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 96994-73-9

H3CNCH3FN

CAS RN 96994-73-9

IUPAC Name: 2-(dimethylamino)-6-fluorobenzonitrile
Synonyms: 2-(dimethylamino)-6-fluorobenzonitrile
Molecular Weight (g/mol): 164.18
Molecular Formula: C9H9FN2
InChi Key: WWNKVUULRIWDJZ-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=CC(F)=C1C#N
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)