accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3CH2OHOH2N

CAS RN 96886-55-4

IUPAC Name: 2-amino-2-methylpent-4-enoic acid
Synonyms: 2-amino-2-methylpent-4-enoic acid
Molecular Weight (g/mol): 129.16
Molecular Formula: C6H11NO2
InChi Key: QMBTZYHBJFPEJB-UHFFFAOYNA-N
SMILES: CC(N)(CC=C)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results