accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 96799-02-9

IUPAC Name: 5-(furan-2-yl)-1H-pyrazol-3-amine
Synonyms: 5-(furan-2-yl)-1H-pyrazol-3-amine
Molecular Weight (g/mol): 149.15
Molecular Formula: C7H7N3O
InChi Key: XJNZHICOWTVWOX-UHFFFAOYSA-N
SMILES: NC1=NNC(=C1)C1=CC=CO1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)