CAS RN 96613-89-7
IUPAC Name:
6-nitrohexane-1,2,3,4,5-pentol hydrate
Synonyms:
6-nitrohexane-1,2,3,4,5-pentol hydrate
Molecular Weight (g/mol):
229.19
Molecular Formula:
C6H15NO8
InChi Key:
OHFQXMNDKUHFDU-UHFFFAOYNA-N
SMILES:
O.OCC(O)C(O)C(O)C(O)C[N+]([O-])=O
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar 1-Deoxy-1-nitro-D-iditol hemihydrate, 99%, Thermo Scientific™
CAS: 96613-89-7 Molecular Formula: C12H28N2O15 Molecular Weight (g/mol): 440.36 MDL Number: MFCD00191971 InChI Key: HQAUHMYCAWVBKQ-UHFFFAOYNA-N Synonym: 1-deoxy-1-nitro-d-iditol hemihydrate,2r,3s,4s,5r-6-nitrohexane-1,2,3,4,5-pentol hydrate PubChem CID: 73995059 IUPAC Name: (2R,3S,4S,5R)-6-nitrohexane-1,2,3,4,5-pentol;hydrate SMILES: O.OCC(O)C(O)C(O)C(O)C[N+]([O-])=O.OCC(O)C(O)C(O)C(O)C[N+]([O-])=O