accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OOOHO

CAS RN 96443-42-4

IUPAC Name: 3-(methoxycarbonyl)cyclopentane-1-carboxylic acid
Synonyms: 3-(methoxycarbonyl)cyclopentane-1-carboxylic acid
Molecular Weight (g/mol): 172.18
Molecular Formula: C8H12O4
InChi Key: FVUHGTQDOMGZOT-UHFFFAOYNA-N
SMILES: COC(=O)C1CCC(C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results