accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OOOOOCH3N

CAS RN 960201-81-4

IUPAC Name: 6,7-dimethyl 8-benzyl-4-oxo-8-azabicyclo[3.2.1]oct-2-ene-6,7-dicarboxylate
Synonyms: 6,7-dimethyl 8-benzyl-4-oxo-8-azabicyclo[3.2.1]oct-2-ene-6,7-dicarboxylate
Molecular Weight (g/mol): 329.35
Molecular Formula: C18H19NO5
InChi Key: PYZQFEIRZQYUJQ-UHFFFAOYNA-N
SMILES: COC(=O)C1C2C=CC(=O)C(C1C(=O)OC)N2CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results