accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OOHNNH2OHO

CAS RN 95753-56-3

IUPAC Name: 3-(4-aminophenyl)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms: 3-(4-aminophenyl)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol): 402.45
Molecular Formula: C24H22N2O4
InChi Key: VALNSJHHRPSUDO-UHFFFAOYNA-N
SMILES: NC1=CC=C(CC(NC(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)C(O)=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results