accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BONHNOHOOHHCl

CAS RN 957060-89-8

IUPAC Name: (3-{[2-(morpholin-4-yl)ethyl]carbamoyl}phenyl)boronic acid hydrochloride
Synonyms: 3-{[2-(morpholin-4-yl)ethyl]carbamoyl}phenylboronic acid hydrochloride
Molecular Weight (g/mol): 314.57
Molecular Formula: C13H20BClN2O4
InChi Key: QBUJRGQOXYFCFK-UHFFFAOYSA-N
SMILES: Cl.OB(O)C1=CC(=CC=C1)C(=O)NCCN1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results