accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BNHONHBOHHOHOOH

CAS RN 957060-87-6

IUPAC Name: [3-({[3-(dihydroxyboranyl)phenyl]carbamoyl}amino)phenyl]boronic acid
Synonyms: 3-({[3-(dihydroxyboranyl)phenyl]carbamoyl}amino)phenylboronic acid
Molecular Weight (g/mol): 299.88
Molecular Formula: C13H14B2N2O5
InChi Key: UQUQNYFFKZOTSS-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(NC(=O)NC2=CC=CC(=C2)B(O)O)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results