CAS RN 956150-13-3
IUPAC Name:
4-(cyclohexa-1,4-dien-1-yl)butan-1-ol
Synonyms:
4-(cyclohexa-1,4-dien-1-yl)butan-1-ol
Molecular Weight (g/mol):
152.24
Molecular Formula:
C10H16O
InChi Key:
QUBPGTNRIJGDFP-UHFFFAOYSA-N
SMILES:
OCCCCC1=CCC=CC1
1
–
1
of
1
results
1
Alfa Aesar™ 4-(1,4-Cyclohexadien-1-yl)-1-butanol, 97%, stab. with 0.1% BHT
CAS: 956150-13-3 Molecular Formula: C10H16O Molecular Weight (g/mol): 152.237 MDL Number: MFCD22682825 InChI Key: QUBPGTNRIJGDFP-UHFFFAOYSA-N Synonym: 4-cyclohexa-1,4-dien-1-yl butan-1-ol,4-1,4-cyclohexadien-1-yl-1-butanol PubChem CID: 57103928 IUPAC Name: 4-cyclohexa-1,4-dien-1-ylbutan-1-ol SMILES: C1C=CCC(=C1)CCCCO