accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 95201-93-7

CH3OHOSBrO(Z)

CAS RN 95201-93-7

IUPAC Name: (2Z)-4-bromo-2-[hydroxy(methoxy)methylidene]-2,3-dihydrothiophen-3-one
Synonyms: (2Z)-4-bromo-2-[hydroxy(methoxy)methylidene]thiophen-3-one
Molecular Weight (g/mol): 237.07
Molecular Formula: C6H5BrO3S
InChi Key: YCTROWLDGFWISA-WAYWQWQTSA-N
SMILES: CO\C(O)=C1/SC=C(Br)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results