accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 95-31-8

IUPAC Name: (1,3-benzothiazol-2-ylsulfanyl)(tert-butyl)amine
Synonyms: (1,3-benzothiazol-2-ylsulfanyl)(tert-butyl)amine
Molecular Weight (g/mol): 238.37
Molecular Formula: C11H14N2S2
InChi Key: IUJLOAKJZQBENM-UHFFFAOYSA-N
SMILES: CC(C)(C)NSC1=NC2=CC=CC=C2S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results