accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 949-90-6

H2NONHOO

CAS RN 949-90-6

IUPAC Name: benzyl N-(carbamoylmethyl)carbamate
Synonyms: benzyl N-(carbamoylmethyl)carbamate
Molecular Weight (g/mol): 208.22
Molecular Formula: C10H12N2O3
InChi Key: HQYMUNCIMNFLDT-UHFFFAOYSA-N
SMILES: NC(=O)CNC(=O)OCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results