CAS RN 948-19-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N,N-Dimethyltryptamine Oxide, TRC
CAS: 948-19-6 Molecular Formula: C12 H16 N2 O Molecular Weight (g/mol): 204.27 Synonym: N,N-Dimethyltryptamine N-oxide,N,N-dimethyl N-oxide-1H-indole-3-ethanamine IUPAC Name: 2-(1H-indol-3-yl)-N,N-dimethyl-ethanamine oxide SMILES: CN(=O)(C)CCc1c[nH]c2ccccc12