accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 945863-21-8

BOOH3CCH3H3CH3CNOHNCH3

CAS RN 945863-21-8

IUPAC Name: N-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carboxamide
Synonyms: N-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carboxamide
Molecular Weight (g/mol): 262.12
Molecular Formula: C13H19BN2O3
InChi Key: LUMBLRWRRONQMO-UHFFFAOYSA-N
SMILES: CNC(=O)C1=NC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results