accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 943319-70-8

CH3ONHNNH3CFFFNNN

CAS RN 943319-70-8

IUPAC Name: 3-(2-{imidazo[1,2-b]pyridazin-3-yl}ethynyl)-4-methyl-N-{4-[(4-methylpiperazin-1-yl)methyl]-3-(trifluoromethyl)phenyl}benzamide
Synonyms: ponatinib
Molecular Weight (g/mol): 532.57
Molecular Formula: C29H27F3N6O
InChi Key: PHXJVRSECIGDHY-UHFFFAOYSA-N
SMILES: CN1CCN(CC2=C(C=C(NC(=O)C3=CC(C#CC4=CN=C5C=CC=NN45)=C(C)C=C3)C=C2)C(F)(F)F)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results