accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 942206-08-8

CH3NNH3COOCH3H3CCH3I

CAS RN 942206-08-8

IUPAC Name: tert-butyl N-(5-iodo-3-methylpyridin-2-yl)-N-methylcarbamate
Synonyms: tert-butyl N-(5-iodo-3-methylpyridin-2-yl)-N-methylcarbamate
Molecular Weight (g/mol): 348.18
Molecular Formula: C12H17IN2O2
InChi Key: HWHYESISLPNYCT-UHFFFAOYSA-N
SMILES: CN(C(=O)OC(C)(C)C)C1=NC=C(I)C=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results