accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NOOONOOONOOOTb

CAS RN 94219-56-4

IUPAC Name: terbium(3+) hydrate trinitrate
Synonyms: terbium(3+) hydrate trinitrate
Molecular Weight (g/mol): 362.95
Molecular Formula: H2N3O10Tb
InChi Key: UNSPFWLYDUXJHT-UHFFFAOYSA-N
SMILES: O.[Tb+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results