accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 941716-96-7

IUPAC Name: 2,3,4,5,6-pentafluorophenyl 1-methyl-1H-indole-7-carboxylate
Synonyms: 2,3,4,5,6-pentafluorophenyl 1-methylindole-7-carboxylate
Molecular Weight (g/mol): 341.24
Molecular Formula: C16H8F5NO2
InChi Key: UEOKRSIJVAQHID-UHFFFAOYSA-N
SMILES: CN1C=CC2=CC=CC(C(=O)OC3=C(F)C(F)=C(F)C(F)=C3F)=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results