accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CNSOOCl

CAS RN 941716-95-6

IUPAC Name: 1-methyl-1H-indole-7-sulfonyl chloride
Synonyms: 1-methylindole-7-sulfonyl chloride
Molecular Weight (g/mol): 229.68
Molecular Formula: C9H8ClNO2S
InChi Key: DVEZDWQOMVECCI-UHFFFAOYSA-N
SMILES: CN1C=CC2=CC=CC(=C12)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results