accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 940362-32-3

NHF

CAS RN 940362-32-3

IUPAC Name: N-[(2-fluorophenyl)methyl]aniline
Synonyms: N-[(2-fluorophenyl)methyl]aniline
Molecular Weight (g/mol): 201.24
Molecular Formula: C13H12FN
InChi Key: HPLOQKBWLYWYHE-UHFFFAOYSA-N
SMILES: FC1=CC=CC=C1CNC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results