accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OOH

CAS RN 938-79-4

IUPAC Name: 2-phenylbutanoic acid
Synonyms: α-phenylbutyric acid α-ethyl-a-toluic acid α-toluic acid, α-ethyl- α-ethylbenzeneacetate α-ethyl-α-toluate α-ethyl-α-toluic acid α-ethylbenzeneacetic acid α-ethyl-a-toluate
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: OFJWFSNDPCAWDK-UHFFFAOYNA-N
SMILES: CCC(C(O)=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (3)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)