accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 938-18-1

CH3CH3H3CClO

CAS RN 938-18-1

IUPAC Name: 2,4,6-trimethylbenzoyl chloride
Synonyms: 2,4,6-trimethylbenzoyl chloride
Molecular Weight (g/mol): 182.65
Molecular Formula: C10H11ClO
InChi Key: UKRQMDIFLKHCRO-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C(Cl)=O)C(C)=C1
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)