accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NNH2NNS

CAS RN 937795-94-3

IUPAC Name: 1-(1-{thieno[3,2-d]pyrimidin-4-yl}piperidin-4-yl)methanamine
Synonyms: 1-(1-{thieno[3,2-d]pyrimidin-4-yl}piperidin-4-yl)methanamine
Molecular Weight (g/mol): 248.35
Molecular Formula: C12H16N4S
InChi Key: ZKNFKYHZGAWSEG-UHFFFAOYSA-N
SMILES: NCC1CCN(CC1)C1=C2SC=CC2=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results