accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BOOH3CCH3H3CH3CNNOOH3CH3CCH3

CAS RN 936694-19-8

IUPAC Name: tert-butyl 4-{[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}piperazine-1-carboxylate
Synonyms: tert-butyl 4-{[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}piperazine-1-carboxylate
Molecular Weight (g/mol): 402.34
Molecular Formula: C22H35BN2O4
InChi Key: AAEYFMAHSYKHGD-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCN(CC2=CC=C(C=C2)B2OC(C)(C)C(C)(C)O2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results