CAS RN 93601-68-4
CAS RN 93601-68-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-O-(Alpha-D-Galactopyranosyl)-D-galactose, TRC
CAS: 93601-68-4 Molecular Formula: C12 H22 O11 Molecular Weight (g/mol): 342.3 IUPAC Name: (3R,4S,5R,6R)-6-(hydroxymethyl)-3-[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxane-2,4,5-triol SMILES: OC[C@H]1OC(O)[C@H](O[C@H]2O[C@H](CO)[C@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@H]1O