accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 934586-50-2

BOOCH3H3CCH3CH3O

CAS RN 934586-50-2

IUPAC Name: 2-(2,3-dihydro-1-benzofuran-7-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(2,3-dihydro-1-benzofuran-7-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 246.11
Molecular Formula: C14H19BO3
InChi Key: HCAAUTYLSSDHFV-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=C2OCCC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results