accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 934570-54-4

IUPAC Name: 4-(3,5-dimethyl-1H-pyrazol-1-yl)benzaldehyde
Synonyms: 4-(3,5-dimethylpyrazol-1-yl)benzaldehyde
Molecular Weight (g/mol): 200.24
Molecular Formula: C12H12N2O
InChi Key: XSNMWJQOHKWNKY-UHFFFAOYSA-N
SMILES: CC1=NN(C(C)=C1)C1=CC=C(C=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results