accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 934570-44-2

IUPAC Name: 3-(1,3-dioxolan-2-yl)thiophene-2-carboxylic acid
Synonyms: 3-(1,3-dioxolan-2-yl)thiophene-2-carboxylic acid
Molecular Weight (g/mol): 200.21
Molecular Formula: C8H8O4S
InChi Key: ZODQJPSGAHBVHU-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(C=CS1)C1OCCO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results