accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 93339-98-1

H3COFOH

CAS RN 93339-98-1

IUPAC Name: 1-(2-fluoro-6-hydroxyphenyl)ethan-1-one
Synonyms: 1-(2-fluoro-6-hydroxyphenyl)ethanone
Molecular Weight (g/mol): 154.14
Molecular Formula: C8H7FO2
InChi Key: PSNPXFMLAVLPPP-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(O)C=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results