CAS RN 932737-65-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
FITM, MedChemExpress
MedChemExpress FITM is a negative allosteric modulator of mGlu1 receptor with a Ki of 2.5 nM.