accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NNHOHNO

CAS RN 932-53-6

IUPAC Name: 6-methyl-2,3,4,5-tetrahydro-1,2,4-triazine-3,5-dione
Synonyms: 6-azathymine
Molecular Weight (g/mol): 127.1
Molecular Formula: C4H5N3O2
InChi Key: XZWMZFQOHTWGQE-UHFFFAOYSA-N
SMILES: CC1=NNC(=O)NC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results