accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OPOHOOHHOOPOOHOOPOOHHOOHOHNH4

CAS RN 93133-76-7

IUPAC Name: ammonium 2,4,5-trihydroxy-3,6-bis(phosphonooxy)cyclohexyl hydrogen phosphate
Synonyms: ammonium 2,4,5-trihydroxy-3,6-bis(phosphonooxy)cyclohexyl hydrogen phosphate
Molecular Weight (g/mol): 437.12
Molecular Formula: C6H18NO15P3
InChi Key: LCBKVESQTQJLKF-UHFFFAOYNA-N
SMILES: [NH4+].OC1C(O)C(OP(O)(O)=O)C(OP(O)([O-])=O)C(O)C1OP(O)(O)=O
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)