accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OH3CNHNH2HCl

CAS RN 93048-16-9

IUPAC Name: (4-methoxy-2-methylphenyl)hydrazine hydrochloride
Synonyms: (4-methoxy-2-methylphenyl)hydrazine hydrochloride
Molecular Weight (g/mol): 188.66
Molecular Formula: C8H13ClN2O
InChi Key: OIFKWIPSJGNUCC-UHFFFAOYSA-N
SMILES: Cl.COC1=CC(C)=C(NN)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results