accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NOHO

CAS RN 930111-13-0

IUPAC Name: {1-[(furan-2-yl)methyl]piperidin-4-yl}methanol
Synonyms: [1-(furan-2-ylmethyl)piperidin-4-yl]methanol
Molecular Weight (g/mol): 195.26
Molecular Formula: C11H17NO2
InChi Key: YBANPRCPQURNFY-UHFFFAOYSA-N
SMILES: OCC1CCN(CC2=CC=CO2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results