accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 930111-07-2

IUPAC Name: methyl({1-[(thiophen-2-yl)methyl]piperidin-4-yl}methyl)amine
Synonyms: methyl({[1-(thiophen-2-ylmethyl)piperidin-4-yl]methyl})amine
Molecular Weight (g/mol): 224.37
Molecular Formula: C12H20N2S
InChi Key: JXFQDIBOGWDEEL-UHFFFAOYSA-N
SMILES: CNCC1CCN(CC2=CC=CS2)CC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results