accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 929-10-2

H3CCH3OOH

CAS RN 929-10-2

IUPAC Name: 6-methylheptanoic acid
Synonyms: cekanoic C8 acid
Molecular Weight (g/mol): 144.21
Molecular Formula: C8H16O2
InChi Key: OEOIWYCWCDBOPA-UHFFFAOYSA-N
SMILES: CC(C)CCCCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results