accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3CH3MgBr

CAS RN 926-62-5

IUPAC Name: magnesium(2+) 2-methylpropan-1-ide bromide
Synonyms: magnesium(2+) 2-methylpropan-1-ide bromide
Molecular Weight (g/mol): 161.33
Molecular Formula: C4H9BrMg
InChi Key: DQDWATOXYCARFV-UHFFFAOYSA-M
SMILES: [Mg++].[Br-].CC(C)[CH2-]
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results